Learn More
Medchemexpress LLC 1-Cbz-3-hydroxyazetidine | 128117-22-6 | MFCD09037849 | 99.9% | 207.23 | C11H13NO3 | 50 G

Supplier: Medchemexpress LLC HY7747550G
1-Cbz-3-hydroxyazetidine is a non-cleavable ADC linker that can also serve as an alkyl chain-based PROTAC linker; it is supplied as a high-purity powder for use in small-molecule conjugation chemistry.
- Purity 99.9%.
- Molecular formula C11H13NO3.
- Molecular weight 207.23.
- CAS number 128117-22-6.
- Available pack size 50 G; larger quantities available by quote.
- Typical storage: powder -20°C for long-term; 4°C for shorter term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.