Learn More
Waters Corp Waters Corporation Collection Plate Spacer Kit, 3 mm, 15 mm, 17 mm Spacers
Supplier:  Waters Corp 186010527
Encompass_Preferred
This kit allows for optimal height placement of collection labware within the Extraction+ device. The spacers can be mixed and matched to achieve different heights and help to avoid cross-contaminations during extraction methods.
  • Allows for optimal height placement of collection labware within the Extraction+ device.
  • Spacers can be mixed and matched to achieve different heights.
  • Helps to avoid cross-contaminations during extraction methods.
  • Includes 2x 3 mm, 1x 15 mm, and 1x 17 mm spacers.
Catalog No. 50-226-9414