Learn More
AdipoGen 1-Chloro-1 2-benziodoxol-3-one
SDP
Supplier:  AdipoGen CDXC0135M100
Encompass_Preferred
Chemical. CAS 59457-26-0. Formula C7H4ClIO2. MW 282.5. Synthetic. A Hypervalent iodine reagent for oxidation reactions.
Catalog No. 50-256-1462