Learn More
Medchemexpress LLC 1-Chloro-2-ethynylbenzene | 873-31-4 | 136.58 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00258610G
Encompass_Preferred
1-Chloro-2-ethynylbenzene is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is a click chemistry reagent, containing an Alkyne group, and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Biochemical reagent for life science related research
  • Biological material or organic compound
  • Click chemistry reagent
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
Catalog No. 50-004-49581