Learn More
Medchemexpress LLC 1-(Chlorocarbonyl)piperidine-d10 | 1219803-31-2 | 98.2% | 157.66 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY33540S50MG
Encompass_Preferred
1-(Chlorocarbonyl)piperidine-d10 is the deuterium labeled 1-(Chlorocarbonyl)piperidine. This compound is intended for research purposes only and is not for patient use. It can serve as a tracer and an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. Deuteration is notable for its potential impact on the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-04454