Learn More
Medchemexpress LLC 1-Chloromethylpyrene 100mg
SDP
Supplier:  Medchemexpress LLC HYW111005100MG
Encompass_Preferred
1-Chloromethylpyrene is an alkylating agent that enters the DNA double helix structure through intercalation forms covalent bonds with electrophilic groups resulting in DNA cross-linking thereby inhibiting the proliferation of tumor cells[1]
Catalog No. 50-002-06370