Learn More
Medchemexpress LLC 1-cyano-2-(2-methylbutan-2-yl)-3-(3-pyridinyl)guanidine | 60559-98-0 | 99.5% | 231.30 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10857350MG
Encompass_Preferred
P-1075 is a potent activator of sulfonylurea receptor 2-associated ATP-sensitive potassium channels (SUR2-KIR6), with an EC50 value of 45 nM for SUR2B-KIR6 channel activation. P-1075 also opens mitochondrial K(ATP) channels and generates reactive oxygen species resulting in cardioprotection of rabbit hearts.
  • Potent activator of sulfonylurea receptor 2-associated ATP-sensitive potassium channels (SUR2-KIR6).
  • EC50 value of 45 nM for SUR2B-KIR6 channel activation.
  • Opens mitochondrial K(ATP) channels.
  • Generates reactive oxygen species.
  • Results in cardioprotection of rabbit hearts.
Catalog No. 50-003-46277