Learn More
Selleck Chemical LLC 1-Cyclohexyl-3-dodecyl urea S0548-5MG
SDP
Supplier:  Selleck Chemical LLC S05485MG
Encompass_Preferred
1-Cyclohexyl-3-dodecyl urea (CDU N-Cyclohexyl-N-dodecyl urea NCND) is a highly selective inhibitor of soluble epoxide hydrolase (sEH) that increases epoxyeicosatrienoic acids (EETs) levels
Catalog No. 50-000-42948