Learn More
Medchemexpress LLC 1-(α-D-ribofuranosyl)uracil | 3258-07-9 | MFCD17430297 | 97.0% | 244.20 g/mol | C9H12N2O6 | 1 ML

Supplier: Medchemexpress LLC HYW12811210MM/1ML
1-(α-D-ribofuranosyl)uracil is a purine nucleoside analog provided as a 10 mM solution in DMSO (1 mL) for biochemical and oncology research. It is reported to inhibit DNA synthesis and induce apoptosis in certain tumor models.
- Supplied as a 10 mM solution in DMSO, 1 mL.
- Molecular formula C9H12N2O6; molecular weight 244.20 g/mol.
- Purity: 97.0%.
- Appearance: white to off-white solid (supplied as solution).
- Storage: in solvent store at -80°C up to 6 months, -20°C up to 1 month.
- Applications: useful for studies of DNA synthesis inhibition and apoptosis.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.