Learn More
Medchemexpress LLC 1-(dibenzylamino)-3-(4-(trifluoromethyl)phenoxy)propan-2-ol | 2222094-18-8 | 99.7% | C24H24F3NO2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13465610MG
Encompass_Preferred
BC1618 is a small-molecule research compound that inhibits Fbxo48 and stimulates AMPK-dependent signaling by preventing pAMPKα degradation. It is used in preclinical studies of mitophagy, mitochondrial dynamics, and metabolic regulation.
  • Orally active tool compound suitable for in vivo studies.
  • Stimulates AMPK-dependent signaling by preventing pAMPKα degradation.
  • Facilitates research into mitophagy, mitochondrial fission, and insulin sensitivity.
  • High reported purity for reliable experimental results.
  • Soluble in DMSO, insoluble in water, enabling formulation flexibility.
  • Available in small quantities suitable for biochemical and animal studies.
Catalog No. 50-002-61527