Learn More
STA PHARMACEUTICAL US LLC 1-DMTr-(S)-2-C9-Glycol amidite | 1 g | InChIKey VDNAYUVRQSPTOJ-VFNQWJAUSA-N | MW 690.905761998
Supplier: STA PHARMACEUTICAL US LLC STAAM107111GM
1-DMTr-(S)-2-C9-Glycol amidite is a Amidite reagent (Subcategory: Spacer Modifiers) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. Ships on dry ice. SDS available for reference.
Specifications
- CAS: No data
- MDL: No data
- InChIKey: VDNAYUVRQSPTOJ-VFNQWJAUSA-N
- Molecular Weight: 690.905761998
- Molecular Formula: C41H59N2O5P
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 7.8 g
- Commodity Code: 29299090
- Country Of Origin: China
- IUPAC: (S)-1-(bis(4-methoxyphenyl)(phenyl)methoxy)undecan-2-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(N(C(C)C)P(OCCC#N)O[C@@H](CCCCCCCCC)COC(C1=CC=C(C=C1)OC)(C2=CC=C(C=C2)OC)C3=CC=CC=C3)C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.