Learn More
Medchemexpress LLC 1-Docosanol (Behenyl alcohol) | 661-19-8 | 98.0% | 1 G

Supplier: Medchemexpress LLC HYB02221G
1-Docosanol (Behenyl alcohol) is a saturated fatty alcohol known for its inhibitory activity against lipid-enveloped viruses, including herpes simplex virus (HSV).
- Inhibits replication of wild-type and acyclovir-resistant HSV in vero cells
- No adverse effects observed in rats during studies
- White to off-white solid appearance
- Derived from plants
- Molecular formula: C22H46O
- Molecular weight: 326.60
- CAS number: 661-19-8
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.