Learn More
Medchemexpress LLC 1-Eicosanol-d41 (Arachidyl alcohol-d41) | 349553-89-5 | 339.80 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW004263S1MG
Encompass_Preferred
1-Eicosanol-d41 (Arachidyl alcohol-d41) is the deuterium-labeled version of 1-Eicosanol, a natural compound with antioxidant activity found in Justicia insularis leaves. It can be used as an antioxidant, tracer, or internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-49108