Learn More
Medchemexpress LLC 1'-epi Gemcitabine hydrochloride | 122111-05-1 | C9H12ClF2N3O4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0003A1MG
Encompass_Preferred
1'-epi Gemcitabine hydrochloride is an isomer of Gemcitabine hydrochloride. It functions as an experimental control. The parent compound, Gemcitabine Hydrochloride, is a pyrimidine nucleoside analog antimetabolite and an antineoplastic agent. It works by inhibiting DNA synthesis and repair, leading to processes such as autophagy and apoptosis.
  • Used as an experimental control
  • Isomer of gemcitabine hydrochloride
  • Parent compound inhibits DNA synthesis and repair
  • Parent compound induces autophagy and apoptosis
Catalog No. 50-004-32545