Learn More
Medchemexpress LLC 1-Ethyl-(3-dimethylaminopropyl)carbodiimide (hydrochloride) | 25952-53-8 | 100.0% | 100 G
SDP
Supplier:  Medchemexpress LLC HYD0178100G
Encompass_Preferred
1-Ethyl-(3-dimethylaminopropyl)carbodiimide hydrochloride is a carbodiimide reagent and aminoaldehyde dehydrogenase (AMADH) inhibitor. It accelerates the formation of esters, amides, and peptides, acting as a condensing and dehydrating agent.
  • Forms nucleic acid and compounds with amide bonds.
  • Used for polynucleotide synthesis, anhydroxydation, lactonization, and esterification.
  • Forms amide bonds (peptide bonds) in aqueous medium.
  • Prepares polyaniline-carbon nanotubes for cholesterol biosensors.
  • Performs precolumn derivatization of aliphatic amines for HPLC.
  • Assists in molecular beacon formation for DNA research.
  • Aids in sensor preparation for calcium detection.
  • Used for fluorescent determination of carboxylic acids.
  • Applied in solid-phase microsequencing of peptides.
Catalog No. 50-004-10157