Learn More
Medchemexpress LLC 1-(Ethyl-d5)-6,8-difluoro-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride | 99.9% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0455S1MG
Encompass_Preferred
Lomefloxacin-d5 hydrochloride is a deuterium labeled version of lomefloxacin hydrochloride, a broad-spectrum quinolone antibiotic with antimicrobial activity. It is utilized in research for various infections, including those of the respiratory tract, genitourinary system, gastrointestinal tract, and ENT.
  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS).
  • Incorporates stable heavy isotopes for quantitation in drug development.
  • Deuteration may affect pharmacokinetic and metabolic drug profiles.
Catalog No. 50-004-05453