Learn More
Cambridge Isotope Laboratories 1% Ethylbenzene + 2% Tms In Chloroform-D
Supplier:  Cambridge Isotope Laboratories DLM64
Encompass
Appearance: Liquid, clear, Molecular mass: 120.38 g/mol, Melting point: -63 °C (-81 °F), Boiling point: 60.5 - 61.5 °C (140.9-142.7 °F), Density: 1.5 g/mL at 25 °C (77 °F).
Catalog No. NC1082078