Learn More
Sigma Aldrich Fine Chemicals Biosciences 1 fluoro 24 dinitrobenzene1G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 7317710X1G
Encompass
1-fluoro-2 4-dinitrobenzene is a color labeling agent mainly used for detecting primary and secondary amines.
Catalog No. 50-289-9408