Learn More
Indofine Chemical 1-Fluoropentane, 98%, 1 Gm
SDP
Supplier:  Indofine Chemical 261004 1 GM
Encompass_Preferred
1-FLUOROPENTANE 1 GM, Fluoro Organics, Amyl fluoride, n-Pentyl fluoride, C5H11F, MW: 90.14, 0.98, 592-50-7, MFCD00000453
Catalog No. 50-911-104