Learn More
US Plastic Corporation 1 GAL JUG WITH CAP 48/CS
SDP
Supplier:  US Plastic Corporation 66158
Encompass
Made of high density polyethylene. Natural color with high corrosion resistance. Similar to jugs used for milk. Built-in handles. Funnel top for easy pouring. Meets FDA standards. Caps are white. Great for packing.
Catalog No. NC9747080