Learn More
Medchemexpress LLC 1-Heptadecanol | 1454-85-9 | 256.47 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW00429650G
Encompass_Preferred
1-Heptadecanol is a long-chain primary alcohol and an antibacterial agent. It can be isolated from *Solena amplexicaulis* and demonstrates antibacterial activity against *Salmonella gallinarum*. It is also used in research related to dandruff caused by *Malassezia furfur*.
  • Long-chain primary alcohol
  • Antibacterial agent
  • Isolated from *Solena amplexicaulis*
  • Exhibits antibacterial activity against *Salmonella gallinarum* (MIC of 15.08 μg/mL)
  • Used in research of dandruff caused by *Malassezia furfur*
Catalog No. 50-004-45416