Learn More
Medchemexpress LLC 1-Heptadecanoyl-2-hydroxy-sn-glycero-3-phosphocholine | 50930-23-9 | MFCD00133433 | 99.0% | C25H52NO7P | 1 MG
Supplier: Medchemexpress LLC HY1416161MG
1-Heptadecanoyl-2-hydroxy-sn-glycero-3-phosphocholine is an endogenous metabolite found in blood. It is used in research related to pregnancy and sepsis. Endogenous metabolites are defined as those annotated by the Kyoto Encyclopedia of Genes and Genomes as substrates or products of the approximately 1900 metabolic enzymes encoded in the human genome.
- Endogenous metabolite present in blood
- Used for research of pregnancy
- Used for research of sepsis
- Purity of 99.0%
- White to off-white solid appearance