Learn More
Chem-Impex International, Inc. 1-Heptanesulfonic acid sodium salt anhydrous , suitable for Ion-Pair Chromatography | 22767-50-6 | MFCD00007543 | 25G
Supplier:  Chem-Impex International, Inc. 2284125G
Encompass_Preferred
1-Heptanesulfonic acid sodium salt anhydrous , suitable for Ion-Pair Chromatography, 22767-50-6, MFCD00007543, 25G
Catalog No. 50-520-298