Learn More
Apexbio Technology LLC 1-Hexadecanol 36653-82-4 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B16515.1
Encompass_Preferred
1-Hexadecanol (CAS 36653-82-4) is a long-chain aliphatic fatty alcohol frequently employed as an intermediate in the synthesis of various surfactants and emollients While it does not directly target specific biological pathways its amphipathic properties enable it to modulate membrane fluidity and influence lipid organization in cellular models In biomedical research 1-hexadecanol is utilized to study membrane biophysics lipid metabolism and as a precursor in the formulation of delivery vehicles for pharmaceuticals Its role in altering membrane properties also makes it valuable for investigating cellular permeability and drug transport mechanisms
Catalog No. 50-000-90604