Learn More
Medchemexpress LLC 1-Hexadecanol | 36653-82-4 | 99.8% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB146510MM/1ML
Encompass_Preferred
1-Hexadecanol is a fatty alcohol and a lipophilic substrate, used for the research of ferroptosis.
  • Increases the level of lipid peroxidation in HT1080 cells.
  • Increases FAR1 KO HT1080 cells sensitivity to ferroptosis.
  • Has no effect on RSL3-induced ferroptosis in HT1080 cells expressing GNPAT or AGPS shRNA.
Catalog No. 50-004-13531