Learn More
Medchemexpress LLC 1-Hexadecanol | 36653-82-4 | MFCD00004760 | 99.1% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYB1465R
Encompass_Preferred
1-Hexadecanol (Standard) is the analytical standard of 1-Hexadecanol. This product is intended for research and analytical applications. It is a fatty alcohol, a lipophilic substrate, and can be used for the research of ferroptosis.
  • Serves as a reference standard for assays
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS
  • Purity of 99.06%
Catalog No. 50-004-14412