Learn More
Medchemexpress LLC 1-hexadecanol-d5 | 1219799-18-4 | 99.2% | 10 MG

Supplier: Medchemexpress LLC HYB1465S210MG
1-Hexadecanol-d5 is the deuterium-labeled analogue of 1-hexadecanol, provided as a high-purity stable isotope standard for analytical use. It is supplied as a white to off-white solid and is intended for use as a tracer or internal standard in quantitative NMR, GC-MS, and LC-MS analyses.
- High purity: 99.20%.
- Molecular formula: C16H29D5O; molecular weight: 247.47.
- Available in small analytical pack sizes, including 10 mg.
- Soluble in DMSO; warming and sonication can improve dissolution.
- Suitable as a tracer and internal standard for quantitative analysis.
- Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
- Appearance: white to off-white solid.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.