Learn More
Medchemexpress LLC 1-Hexadecanol-d5 | 1219799-18-4 | 99.2% | 5 MG
Supplier: Medchemexpress LLC HYB1465S25MG
1-Hexadecanol-d5 is the deuterium labeled 1-Hexadecanol, which is a fatty alcohol and a lipophilic substrate. This compound is suitable for use as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. The deuteration of this compound is notable for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
- Used as a tracer.
- Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
- Deuteration can influence the pharmacokinetic and metabolic profiles of drugs.