Learn More
Medchemexpress LLC 1-Hexadecanol (Palmityl alcohol) | 36653-82-4 | MFCD00004760 | 99.8% | 50 G
SDP
Supplier:  Medchemexpress LLC HYB146550G
Encompass_Preferred
1-Hexadecanol is a fatty alcohol and a lipophilic substrate that can be used for the research of ferroptosis. It is an endogenous metabolite with various effects on cell lines related to ferroptosis.
  • Endogenous metabolite
  • Can be used for the research of ferroptosis
  • Increases the level of lipid peroxidation in HT1080 cells at 20 μM for 12 hours
  • Increases FAR1 KO HT1080 cells sensitivity to ferroptosis at 10 μM for 12 hours
  • Has no effect on RSL3-induced ferroptosis in HT1080 cells expressing GNPAT or AGPS shRNA at 20 μM for 12 hours
Catalog No. 50-004-12689