Learn More
Medchemexpress LLC 1-Hexadecanol (Standard) | 36653-82-4 | MFCD00004760 | 99.1% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB1465R500MG
Encompass_Preferred
1-Hexadecanol (Standard), also known as Palmityl alcohol (Standard), is an analytical standard of 1-Hexadecanol with a purity of 99.06%. This product is designed for research and analytical applications, functioning as a fatty alcohol and a lipophilic substrate. It is specifically applicable in the research of ferroptosis and serves as a reference standard for various analytical techniques, including HPLC, GC, and MS, across qualitative, quantitative, and methodological studies.
  • Analytical standard of 1-Hexadecanol.
  • Purity: 99.06%.
  • Functions as a fatty alcohol and a lipophilic substrate.
  • Can be used for the research of ferroptosis.
  • Serves as a reference standard for HPLC, GC, and MS applications.
Catalog No. 50-004-12639