Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (±)-1-Hexadecanoyl-3-chloropropane-dd5-diol | 1189718-15-7 | C19H32D5ClO3 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY151895S5MG

Encompass_Preferred

(±)-1-Hexadecanoyl-3-chloropropane-dd5-diol is a deuterium-labeled compound, specifically a (±)-1-Hexadecanoyl-3-chloropropane-diol derivative. Deuterium, a stable heavy isotope of hydrogen, is incorporated into drug molecules as tracers for quantitation during drug development. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs. This product has been tested and complies with specifications, intended for research use only.

  • Appearance: White to off-white liquid
  • Purity (HPLC): 98.1%
  • Isotopic enrichment: 98.7%
  • Molecular weight: 353.98

Catalog No. 50-003-73926


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.