Learn More
Medchemexpress LLC 1-hexanol-d11 | 2159-18-4 | 99.3% | 113.24 g/mol | C6H3D11O | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW032022S1100MG
Encompass_Preferred
1-Hexanol-d11 is the deuterium-labeled analogue of 1-hexanol used as an isotopic tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. Supplied as a liquid (colorless to light yellow) with high isotopic and chemical purity, it is intended for analytical and research applications requiring mass or NMR differentiation.
  • Deuterium-labeled internal standard for accurate quantification.
  • Compatible with NMR, GC-MS, and LC-MS workflows.
  • High chemical purity for reliable analytical results.
  • Available in multiple small package sizes for laboratory use.
  • Liquid form simplifies spiking and sample preparation.
  • Suitable as a tracer in analytical method development.
Catalog No. 50-002-99853