Learn More
Medchemexpress LLC 1-hexanol-d5 | 64118-18-9 | 98.0% | 107.21 | C6H9D5O | 10 MG

Supplier: Medchemexpress LLC HYW032022S310MG
1-Hexanol-d5 is the deuterium-labeled analogue of 1-hexanol provided as a small-quantity analytical standard for use as an internal standard or tracer in mass spectrometry and other analytical workflows. It supports accurate quantitation and method development where an isotopically labeled reference is required.
- Deuterium-labeled internal standard for GC-MS and LC-MS.
- Molecular formula C6H9D5O and molecular weight 107.21.
- CAS number 64118-18-9 for unambiguous identification.
- Available in small pack sizes suitable for analytical applications.
- Suitable as an isotopic tracer and internal reference in quantitative assays.
- Consult the batch certificate of analysis for purity and storage conditions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.