Learn More
BIOGEMS INTERNATIONAL INC 1 Human IL-10 Pre-Coated ELISA Kit
SDP
Supplier:  BIOGEMS INTERNATIONAL INC BGK223011KIT
Encompass
IL-10 is an immunosuppressive cytokine produced by a variety of mammalian cell types including macrophages, monocytes, T cells, B cells and keratinocytes. IL-10 inhibits the expression of proinflammatory cytokines such as IL-1 and TNF-α. Like IL-10, IL-10 enhances humoral immune responses and attenuates cell-mediated immune reactions. Human IL-10 is active on murine cells, but murine IL-10 is inactive on human cells.The BioGems Human IL-10 Pre-Coated ELISA (Enzyme-Linked Immunosorbent Assay) kit is a solid phase immunoassay specially designed to measure Human IL-10 with a 96-well strip plate that is pre-coated with mouse monoclonal antibody specific for IL-10. It contains E. coli-expressed recombinant Human IL-10. The detection antibody is a biotinylated goat polyclonal antibody specific for IL-10. The kit is analytically validated with ready to use reagents.
Catalog No. 50-153-0594