Learn More
Medchemexpress LLC 1α-hydroxy-3-epi-vitamin D3 | 58028-00-5 | 99.3% | 400.64 | C27H44O2 | 5 MG

Supplier: Medchemexpress LLC HY10003A5MG
1α-Hydroxy-3-epi-vitamin D3 is a research-grade metabolite of 1α,25-dihydroxyvitamin D3 that acts as a potent suppressor of parathyroid hormone (PTH) secretion. It is provided as a highly pure compound with defined physicochemical properties for biochemical and pharmacological studies; handle and store according to supplier guidance.
- Natural metabolite of 1α,25-dihydroxyvitamin D3.
- Potent suppressor of parathyroid hormone secretion.
- High purity (99.3%) suitable for research applications.
- Molecular formula C27H44O2; molecular weight 400.64.
- Soluble in DMSO (50 mg/mL); store sealed, away from moisture and light.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.