Learn More
Medchemexpress LLC 1-hydroxy-7-azabenzotriazole | 39968-33-7 | MFCD00210053 | 25g
SDP
Supplier:  Medchemexpress LLC HYZ027525G
Encompass_Preferred
HOAT is a commonly used peptide bond forming agent that may become an impurity in drug synthesis but is not mutagenic HOAT can be used for peptide fragment polycondensation and can also inhibit the racemization of peptide polycondensation[1]
Catalog No. 50-002-66115