Learn More
Medchemexpress LLC 1-Isothiocyanato-PEG4-alcohol | 1835759-69-7 | MFCD29052173 | C9H17NO4S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14124650MG
Encompass_Preferred
1-Isothiocyanato-PEG4-alcohol is a PEG-based PROTAC linker. It is intended for research use only, and not for sale to patients. PROTACs consist of two distinct ligands connected by a linker, where one ligand binds to an E3 ubiquitin ligase and the other to the target protein. PROTACs leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-55821