Learn More
New England Biolabs, Inc. 1 kb Plus DNA Ladder for Safe Stains – 1.25 ml
SDP
Supplier:  New England Biolabs, Inc. N0559S
Encompass
1 kb Plus DNA Ladder for Safe Stains is a pre-mixed, ready-to-load molecular weight marker containing one tracking dye. It has been optimized for use on GelRed, GelGreen and SYBR Safe precast agarose gels. 1 kb Plus DNA Ladder for Safe Stains consists of proprietary plasmids, which are digested to completion with appropriate restriction enzymes to yield 19 bands suitable for use as molecular weight standards for agarose gel electrophoresis. The digested DNA includes fragments ranging from 100 bp to 10 kb. The 0.5, 1.0 and 3.0 kb bands have increased intensity to serve as reference bands.
Catalog No. 50-195-9411