Learn More
Research Products International Corp 1 L Flask Clamp for use with Mini-300 Orbital-Genie
SDP
Supplier:  Research Products International Corp 400842
Encompass
Flask Clamps are for use with the Mini-300 Orbital-Genie and are made of stainless steel for added strength and safety. Mix and match different size clamps on same platform or use all same size. Includes mounting screws.
Note: Flask Clamps can also be used on the Orbital-Genie model with Adhering Mat by removing the blue Adhering Mat.
Catalog No. 50-197-8144