Learn More
SKC Inc SamplePro FlexFilm 1-liter Sample Bag, 10/pack
SDP
Supplier:  SKC Inc 236001
Encompass
SamplePro FlexFilm Air Sample Bags offer good stability for VOCs, carbon monoxide, carbon dioxide, methane, sulfur hexafluoride, and some sulfur compounds. Use PTFE tubing to attach a single polypropylene or dual stainless steel fitting on the bag to a Grab Air or Pocket Pump TOUCH.
Catalog No. NC0293602