Learn More
Medtec Medical Inc 1 LEG GRAVITY FILL SET 50/CS
SDP
Supplier:  Medtec Medical Inc 58794
Encompass
1 Leg Gravity Fill Set, 50/CSDetachable Gravity Fill Legs with female screw connectorFor Secure™ Legless bags with male screw connector Latex freeAvailable in 1, 2 or 3 leg versionsFill legs have vented spike
Catalog No. NC2959551