Learn More
Apexbio Technology LLC 1-Methoxy PMS(Synonyms: 1-Methoxyphenazine methosulfate, 1-Methoxy PMS, Methoxyphenazine methosulfate, MPMS), 200mg, CAS: 65162-13-2.
SDP
Supplier:  Apexbio Technology LLC B8597200
Encompass_Preferred
1-Methoxy PMS (CAS 65162-13-2) is a redox-active small molecule utilized as an electron transfer mediator in biochemical assays Acting at the interface between NAD(P)H and tetrazolium salts 1-Methoxy PMS facilitates the controlled generation of reactive oxygen species (ROS) thereby modulating oxidative stress conditions Due to its ability to regulate cellular redox balance it is widely employed in research exploring oxidative signaling pathways ROS-mediated cellular responses and related pathophysiological processes
Catalog No. 50-000-60745