Learn More
Medchemexpress LLC 1-Methoxynaphthalene | 2216-69-5 | 98.6% | 158.20 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW012568
Encompass_Preferred
1-Methoxynaphthalene is used as a substrate to investigate the activity of cytochrome c peroxidase (CcP). It can also be used to synthesize prenyl naphthalen-ols.
  • Used as a substrate to investigate the activity of cytochrome c peroxidase (CcP).
  • Can be used to synthesize prenyl naphthalen-ols.
Catalog No. 50-004-51217