Learn More
Medchemexpress LLC 1-Methoxyphaseollidin | 65428-13-9 | 1 MG

Supplier: Medchemexpress LLC HYN84891MG
1-Methoxyphaseollidin is a flavonoid compound that acts as a lysoPAF acetyltransferase inhibitor with an IC50 of 48 μM. It demonstrates anti-H.pylori activity against CLAR and AMOX-resistant strains, as well as four CLAR (AMOX)-sensitive strains. This product is for research use only and not sold to patients.
- Flavonoid compound
- LysoPAF acetyltransferase inhibitor (IC50 of 48 μM)
- Exhibits anti-H.pylori activity
- Effective against CLAR and AMOX-resistant H.pylori strains
- Also effective against four CLAR (AMOX)-sensitive strains
- Molecular weight: 354.40
- Formula: C21H22O5
- Appearance: solid
- Color: white to off-white
- Initial source: plants (Leguminosae, Glycyrrhiza uralensis Fisch.)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.