Learn More
Medchemexpress LLC 1-methyl-1,4-dihydronicotinamide | 17750-23-1 | MFCD006277282 | >98.0% | 138.17 g/mol | C7H10N2O | 5MG
SDP
Supplier:  Medchemexpress LLC HY1243135MG
Encompass_Preferred
1-Methyl-1,4-dihydronicotinamide (CAS 17750-23-1) is a reduced nicotinamide derivative used in biochemical research as a radical scavenger and an NADH model compound. It is typically supplied as a crystalline solid of high purity and is used as a substrate or inhibitor in studies of NADH-dependent enzymes and serine racemase activity.
  • Used as a radical scavenger in redox and oxidative stress studies.
  • Serves as an NADH model and substrate for enzyme assays.
  • High reported purity suitable for research applications.
  • Supplied as a crystalline solid with typical cold-storage recommendations.
  • Available in small-mass vials for laboratory assays and screening.
Catalog No. 50-002-38555