Learn More
Medchemexpress LLC 1-Methyl-1H-indazole-3-carboxylic acid | 50890-83-0 | 176.17 | 50 G

Supplier: Medchemexpress LLC HYW00792750G
1-Methyl-1H-indazole-3-carboxylic acid is a versatile biochemical reagent ideal for life science research. It functions as both a biological material and an organic compound, presenting as a white to off-white solid.
- Suitable for life science-related research
- Can be utilized as a biological material or organic compound
- Presents as a white to off-white solid
- Molecular weight of 176.17
- Soluble in DMSO at concentrations of 100 mg/mL or higher (567.63 mM)
- Storage recommendations: 3 years at room temperature; in solvent, 2 years at -80°C, or 1 year at -20°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.