Learn More
Medchemexpress LLC 1-methyl-1H-pyrazole-4-boronic acid pinacol ester-d3 | 1022151-50-3 | ≥98.0% | 211.08 | C10H14D3BN2O2 | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW249514250MG
Encompass_Preferred
Deuterium-labeled boronic ester reagent used as a synthetic building block and analytical standard for research applications. Supplied as a high-purity solid suitable for incorporation in organic synthesis and isotopic labeling studies, and available in small laboratory-scale packages.
  • Deuterium-labeled for isotopic tracing and mass spectrometry studies.
  • High purity: ≥98.0%.
  • Suitable as a boronic ester building block in cross-coupling and synthesis.
  • Offered in small laboratory-scale packages (e.g., 250 mg).
Catalog No. 50-002-80860