Learn More
Medchemexpress LLC 1-Methyl-2-pentyl-4(1H)-quinolinone | 22048-98-2 | 229.32 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN16375MG
Encompass_Preferred
1-Methyl-2-pentyl-4(1H)-quinolinone is a quinolone alkaloid isolated from the fruits of Evodia Rutaecarpa. It possesses antibacterial and cytotoxic activities for cancer cells.
  • Quinolone alkaloid
  • Isolated from the fruits of Evodia Rutaecarpa
  • Possesses antibacterial activities
  • Exhibits cytotoxic activities for cancer cells
Catalog No. 50-004-16606