Learn More
Medchemexpress LLC 1-methyl-2-pyrrolidinone-d9 | 185964-60-7 | MFCD00148961 | 98.0% | 108.19 | C5D9NO | 25mg
SDP
Supplier:  Medchemexpress LLC HYY1275S25MG
Encompass_Preferred
N-Methylpyrrolidone-d9 (NMP-d9) is deuterium labeled N-Methylpyrrolidone N-Methylpyrrolidone (1-Methyl-2-pyrrolidinone) a five-membered cyclic amide is an orally active organic polar solvent with teratogenicity and toxicity N-Methylpyrrolidone is low in acute toxicity with a LD50 value of 3914 mg/kg in rats and of 4050 mg/kg in mice N-Methylpyrrolidone is extensively used in the manufacture of adhesives paints fuels and pharmaceuticals[1][2][3][4]
Catalog No. 50-002-66866