Learn More
Sigma Aldrich Fine Chemicals Biosciences 1-Methyl-2-pyrrolidinone s1L
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 2704581L
Encompass
1-Methyl-2-pyrrolidinone, a tertiary amide, is widely employed as an organic solvent. It participates in the sonochemical synthesis of high quality MOF(Metal-Organic Framework)-177 crystals.It also participates in the followingsynthesis of cyanotrimethylsilanedecarbonylation of aldehydesreduction of benzaldehydes and ketones to the corresponding alcoholspreparation of natural products
Catalog No. 50-289-1470